Methyl 3-(methylcarbamoyl)benzoate 96%
Dodavatel: Apollo Scientific
OR908023-1G
OR908023-5G
APOSOR908023-1GEA
4570
CZK
APOSOR908023-1G
APOSOR908023-5G
Methyl 3-(methylcarbamoyl)benzoate 96%
Methyl 3-(methylcarbamoyl)benzoate
Vzorec:
C₁₀H₁₁NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09234741 CAS číslo: 23668-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...