Tisk…

4-(Ethylcarbamoyl)benzoic acid 98%

Dodavatel: Apollo Scientific

OR907989-1G OR907989-5G
APOSOR907989-1GEA 5200 CZK
APOSOR907989-1G APOSOR907989-5G
4-(Ethylcarbamoyl)benzoic acid 98%
4-(Ethylcarbamoyl)benzoic acid
Vzorec: C₁₀H₁₁NO₃
MW: 193,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12173485
CAS číslo: 167627-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR