Methyl 3-(2-bromoacetyl)benzoate 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR907935-1G
OR907935-5G
APOSOR907935-1GEA
3810
CZK
APOSOR907935-1G
APOSOR907935-5G
Methyl 3-(2-bromoacetyl)benzoate 96%
Methyl 3-(2-bromoacetyl)benzoate
Vzorec:
C₁₀H₉BrO₃ MW: 257.081 g/mol Bod tání: 68…70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09834617 CAS číslo: 27475-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...