Tisk…

4-(Acetamidomethyl)benzoic acid 98%

Dodavatel: Apollo Scientific

OR907934-1G OR907934-5G
APOSOR907934-1GEA 7670 CZK
APOSOR907934-1G APOSOR907934-5G
4-(Acetamidomethyl)benzoic acid 98%
4-(Acetamidomethyl)benzoic acid
Vzorec: C₁₀H₁₁NO₃
MW: 193.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655692
CAS číslo: 1205-58-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR