Tisk…

1,3-benzoxazole-5-carboxylic acid 98%

Dodavatel: Apollo Scientific

Synonyms: Benzo[d]oxazole-5-carboxylic acid

OR907912-1G OR907912-250MG
APOSOR907912-1GEA 7080 CZK
APOSOR907912-1G APOSOR907912-250MG
1,3-benzoxazole-5-carboxylic acid 98%
1,3-benzoxazole-5-carboxylic acid
Vzorec: C₈H₅NO₃
MW: 163,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954950
CAS číslo: 15112-41-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR