Tisk…

3-Hydroxynaphthalene-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Hydroxy-2-naphthaldehyde

OR907899-100MG OR907899-1G OR907899-250MG
APOSOR907899-100MGEA 2480 CZK
APOSOR907899-100MG APOSOR907899-1G APOSOR907899-250MG
3-Hydroxynaphthalene-2-carbaldehyde 95%
3-Hydroxynaphthalene-2-carbaldehyde
Vzorec: C₁₁H₈O₂
MW: 172,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079346
CAS číslo: 581-71-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR