Tisk…

tert-butyl thiophene-2-carboxylate 98%

Dodavatel: Apollo Scientific

OR907891-1G OR907891-5G
APOSOR907891-1GEA 5200 CZK
APOSOR907891-1G APOSOR907891-5G
tert-butyl thiophene-2-carboxylate 98%
tert-butyl thiophene-2-carboxylate
Vzorec: C₉H₁₂O₂S
MW: 184,255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18909502
CAS číslo: 939-62-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR