Tisk…

2-Isopropoxy-2-phenylacetic acid 97%

Dodavatel: Apollo Scientific

OR907836-1G OR907836-250MG
APOSOR907836-1GEA 11380 CZK
APOSOR907836-1G APOSOR907836-250MG
2-Isopropoxy-2-phenylacetic acid 97%
2-Isopropoxy-2-phenylacetic acid
Vzorec: C₁₁H₁₄O₃
MW: 194,227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16661867
CAS číslo: 5394-87-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR