Tisk…

(E)-3-Iodoacrylic acid 97%

Dodavatel: Apollo Scientific

OR907662-10G OR907662-1G OR907662-5G
APOSOR907662-10GEA 28540 CZK
APOSOR907662-10G APOSOR907662-1G APOSOR907662-5G
(E)-3-Iodoacrylic acid 97%
(E)-3-Iodoacrylic acid
Vzorec: C₃H₃IO₂
MW: 197,959 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265244
CAS číslo: 6372-02-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR