Tisk…

Ethyl 3-cyanobenzoate 98%

Dodavatel: Apollo Scientific

OR907617-1G OR907617-5G
APOSOR907617-1GEA 870 CZK
APOSOR907617-1G APOSOR907617-5G
Ethyl 3-cyanobenzoate 98%
Ethyl 3-cyanobenzoate
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03086010
CAS číslo: 2463-16-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR