Tisk…

Methyl 2-chloro-4-methylbenzoate 98%

Dodavatel: Apollo Scientific

OR907520-1G OR907520-5G
APOSOR907520-1GEA 662 CZK
APOSOR907520-1G APOSOR907520-5G
Methyl 2-chloro-4-methylbenzoate 98%
Methyl 2-chloro-4-methylbenzoate
Vzorec: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398994
CAS číslo: 195318-63-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR