Tisk…

2-(2-Methoxyethoxy)pyridine-5-boronic acid 96%

Dodavatel: Apollo Scientific

Synonyms: 2-(2-Methoxyethoxy)-pyridine-5-boronic acid, (6-(2-Methoxyethoxy)pyridin-3-yl)boronic acid

OR907503-1G OR907503-5G
APOSOR907503-1GEA 4070 CZK
APOSOR907503-1G APOSOR907503-5G
2-(2-Methoxyethoxy)pyridine-5-boronic acid 96%
2-(2-Methoxyethoxy)pyridine-5-boronic acid
Vzorec: C₈H₁₂BNO₄
MW: 196,996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045258
CAS číslo: 1282378-03-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR