Tisk…

Cyclohexylmethylboronic acid 98%

Dodavatel: Apollo Scientific

OR907478-1G OR907478-5G
APOSOR907478-1GEA 1700 CZK
APOSOR907478-1G APOSOR907478-5G
Cyclohexylmethylboronic acid 98%
Cyclohexylmethylboronic acid
Vzorec: C₇H₁₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD09878538
CAS číslo: 27762-64-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR