Tisk…

3-Nitrophenethyl alcohol 97%

Dodavatel: Apollo Scientific

Synonyms: 2-(3-Nitrophenyl)ethanol

OR907330-1G OR907330-5G
APOSOR907330-1GEA 2980 CZK
APOSOR907330-1G APOSOR907330-5G
3-Nitrophenethyl alcohol 97%
3-Nitrophenethyl alcohol
Vzorec: C₈H₉NO₃
MW: 167,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010445
CAS číslo: 52022-77-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR