Tisk…

2-Ethoxy-3-methylpyridine-5-boronic acid 98%

Dodavatel: Apollo Scientific

Synonyms: (6-Ethoxy-5-methylpyridin-3-yl)boronic acid

OR907323-1G OR907323-5G
APOSOR907323-1GEA 3610 CZK
APOSOR907323-1G APOSOR907323-5G
2-Ethoxy-3-methylpyridine-5-boronic acid 98%
2-Ethoxy-3-methylpyridine-5-boronic acid
Vzorec: C₈H₁₂BNO₃
MW: 180,997 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181608
CAS číslo: 1451391-77-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR