2-(3-Chlorophenyl)propan-2-ol 95%
Dodavatel: Apollo Scientific
OR907262-1G
OR907262-5G
APOSOR907262-1GEA
6020
CZK
APOSOR907262-1G
APOSOR907262-5G
2-(3-Chlorophenyl)propan-2-ol 95%
2-(3-Chlorophenyl)propan-2-ol
Vzorec:
C₉H₁₁ClO MW: 170,636 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00229409 CAS číslo: 31002-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...