[1-(Butylamino)-1-methyl-ethyl]phosphinic acid 98%
Dodavatel: Apollo Scientific
OR907240-1G
OR907240-5G
APOSOR907240-1GEA
7670
CZK
APOSOR907240-1G
APOSOR907240-5G
[1-(Butylamino)-1-methyl-ethyl]phosphinic acid 98%
[1-(Butylamino)-1-methyl-ethyl]phosphinic acid
Vzorec:
C₇H₁₈NO₂P Storage Temperature: Ambient |
MDL Number:
MFCD00869071 CAS číslo: 17316-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...