Tisk…

5-Methyl-2-propyl-2H-pyrazol-3-ylamine 95%

Dodavatel: Apollo Scientific

OR906963-1G OR906963-5G
APOSOR906963-1GEA 8270 CZK
APOSOR906963-1G APOSOR906963-5G
5-Methyl-2-propyl-2H-pyrazol-3-ylamine 95%
5-Methyl-2-propyl-2H-pyrazol-3-ylamine
Vzorec: C₇H₁₃N₃
Storage Temperature: Ambient
MDL Number: MFCD05237221
CAS číslo: 3524-34-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR