Tisk…

5-Cyano-2-hydroxypyrimidine 95%

Dodavatel: Apollo Scientific

OR906931-1G OR906931-250MG
APOSOR906931-1GEA 16340 CZK
APOSOR906931-1G APOSOR906931-250MG
5-Cyano-2-hydroxypyrimidine 95%
5-Cyano-2-hydroxypyrimidine
Vzorec: C₅H₃N₃O
MW: 121,097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109807
CAS číslo: 1753-49-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR