Tisk…
o-Cyanophenylethylamine hydrochloride
Dodavatel: Apollo Scientific
OR906734-1G OR906734-250MG
APOSOR906734-1GEA 34060 CZK
APOSOR906734-1G APOSOR906734-250MG
o-Cyanophenylethylamine hydrochloride
o-Cyanophenylethylamine hydrochloride
Vzorec: C₉H₁₀N₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD09878804
CAS číslo: 1159826-34-2