Tisk…
1,1-Cyclopropane dimethanol cyclic sulphite 98%
Dodavatel: Apollo Scientific
Synonyms: 5,7-Dioxa-6-thiaspiro[2.5]octane 6-oxide
OR906581-25G OR906581-5G
APOSOR906581-25GEA 36700 CZK
APOSOR906581-25G APOSOR906581-5G
1,1-Cyclopropane dimethanol cyclic sulphite 98%
1,1-Cyclopropane dimethanol cyclic sulphite
Vzorec: C₅H₈O₃S
MW: 148,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832878
CAS číslo: 89729-09-9