Tisk…

Methyl 2,4,6-trimethylbenzoate 95%

Dodavatel: Apollo Scientific

OR906550-1G OR906550-5G
APOSOR906550-1GEA 6220 CZK
APOSOR906550-1G APOSOR906550-5G
Methyl 2,4,6-trimethylbenzoate 95%
Methyl 2,4,6-trimethylbenzoate
Vzorec: C₁₁H₁₄O₂
MW: 178,228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00184961
CAS číslo: 2282-84-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR