Tisk…

1-Methyl-4-(2-pyrrolidinyl)piperidine 95%

Dodavatel: Apollo Scientific

OR906507-1G OR906507-250MG
APOSOR906507-1GEA 9920 CZK
APOSOR906507-1G APOSOR906507-250MG
1-Methyl-4-(2-pyrrolidinyl)piperidine 95%
1-Methyl-4-(2-pyrrolidinyl)piperidine
Vzorec: C₁₀H₂₀N₂
MW: 168,279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05189286
CAS číslo: 524674-26-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR