2-(5-Methylthiophen-2-yl)acetic acid 95%
Dodavatel: Apollo Scientific
OR906497-1G
OR906497-250MG
APOSOR906497-1GEA
24600
CZK
APOSOR906497-1G
APOSOR906497-250MG
2-(5-Methylthiophen-2-yl)acetic acid 95%
2-(5-Methylthiophen-2-yl)acetic acid
Vzorec:
C₇H₈O₂S MW: 156,202 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02664815 CAS číslo: 70624-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...