Tisk…

N'-Benzyl-N'-methyl-ethane-1,2-diamine 97%

Dodavatel: Apollo Scientific

OR906489-1G OR906489-5G
APOSOR906489-1GEA 3740 CZK
APOSOR906489-1G APOSOR906489-5G
N'-Benzyl-N'-methyl-ethane-1,2-diamine 97%
N'-Benzyl-N'-methyl-ethane-1,2-diamine
Vzorec: C₁₀H₁₆N₂
Storage Temperature: Ambient
MDL Number: MFCD03030356
CAS číslo: 14165-18-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR