(2S)-2-(2-Formyl-1H-pyrrol-1-yl)propanoic acid 93%
Dodavatel: Apollo Scientific
OR906223-100MG
OR906223-1G
OR906223-250MG
APOSOR906223-100MGEA
4140
CZK
APOSOR906223-100MG
APOSOR906223-1G
APOSOR906223-250MG
(2S)-2-(2-Formyl-1H-pyrrol-1-yl)propanoic acid 93%
(2S)-2-(2-Formyl-1H-pyrrol-1-yl)propanoic acid
Vzorec:
C₈H₉NO₃ MW: 167,162 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832437 CAS číslo: 145041-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...