(4-Phenyl-1,3-thiazol-2-yl)methanamine 95%
Dodavatel: Apollo Scientific
OR906221-1G
OR906221-250MG
OR906221-5G
APOSOR906221-1GEA
5430
CZK
APOSOR906221-1G
APOSOR906221-250MG
APOSOR906221-5G
(4-Phenyl-1,3-thiazol-2-yl)methanamine 95%
(4-Phenyl-1,3-thiazol-2-yl)methanamine
Vzorec:
C₁₀H₁₀N₂S Storage Temperature: Ambient |
MDL Number:
MFCD05221867 CAS číslo: 90916-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...