Spiro[2.5]octan-6-ylmethanol 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR906089-100MG
OR906089-250MG
APOSOR906089-100MGEA
3060
CZK
APOSOR906089-100MG
APOSOR906089-250MG
Spiro[2.5]octan-6-ylmethanol 97%
Spiro[2.5]octan-6-ylmethanol
Vzorec:
C₉H₁₆O Storage Temperature: Ambient |
MDL Number:
MFCD11501505 CAS číslo: 849671-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...