Monomethyl 3-methylglutarate 98%
Dodavatel: Apollo Scientific
Synonyms:
5-Methoxy-3-methyl-5-oxopentanoic acid
OR905962-100G
OR905962-25G
APOSOR905962-100GEA
11580
CZK
APOSOR905962-100G
APOSOR905962-25G
Monomethyl 3-methylglutarate 98%
Monomethyl 3-methylglutarate
Vzorec:
C₇H₁₂O₄ MW: 160,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631203 CAS číslo: 27151-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...