Tisk…

5-Ethyl-4H-[1,2,4]triazol-3-ylamine 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Amino-5-ethyl-1,2,4-triazole, 5-Ethyl-1H-1,2,4-triazol-3-amine, 5-Ethyl-4H-1,2,4-triazol-3-amine

OR905740-1G OR905740-5G
APOSOR905740-1GEA 2900 CZK
APOSOR905740-1G APOSOR905740-5G
5-Ethyl-4H-[1,2,4]triazol-3-ylamine 97%
5-Ethyl-4H-[1,2,4]triazol-3-ylamine
Vzorec: C₄H₈N₄
Storage Temperature: Ambient
MDL Number: MFCD03085910
CAS číslo: 22819-05-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR