Tisk…
Ethyl (2-formyl-1H-pyrrol-1-yl)acetate 95%
Dodavatel: Apollo Scientific
Synonyms: Ethyl 2-(2-formyl-1H-pyrrol-1-yl)acetate
OR905564-1G OR905564-250MG
APOSOR905564-1GEA 8040 CZK
APOSOR905564-1G APOSOR905564-250MG
Ethyl (2-formyl-1H-pyrrol-1-yl)acetate 95%
Ethyl (2-formyl-1H-pyrrol-1-yl)acetate
Vzorec: C₉H₁₁NO₃
MW: 181,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08277229
CAS číslo: 70582-73-9