Tisk…

Diphenylmethyl isocyanide 95%

Dodavatel: Apollo Scientific

OR905550-100MG OR905550-1G OR905550-250MG
APOSOR905550-100MGEA 5390 CZK
APOSOR905550-100MG APOSOR905550-1G APOSOR905550-250MG
Diphenylmethyl isocyanide 95%
Diphenylmethyl isocyanide
Vzorec: C₁₄H₁₁N
MW: 193,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664566
CAS číslo: 3128-85-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR