Tisk…

1-(3-Chloro-4-methylphenyl)-1H-pyrrole 95%

Dodavatel: Apollo Scientific

OR905315-1G OR905315-250MG
APOSOR905315-1GEA 19440 CZK
APOSOR905315-1G APOSOR905315-250MG
1-(3-Chloro-4-methylphenyl)-1H-pyrrole 95%
1-(3-Chloro-4-methylphenyl)-1H-pyrrole
Vzorec: C₁₁H₁₀ClN
MW: 191,657 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02665246
CAS číslo: 142044-87-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR