Tisk…

(2R,3S,4R,5S)-1-(But-1-yl)-2-(hydroxymethyl)piperidine-3,4,5-triol

Dodavatel: Apollo Scientific
Varování

OR9050T-5MG OR9050T-1MG OR9050T-25MG
APOSOR9050T-5MGEA 11810 CZK
APOSOR9050T-5MG APOSOR9050T-1MG APOSOR9050T-25MG
(2R,3S,4R,5S)-1-(But-1-yl)-2-(hydroxymethyl)piperidine-3,4,5-triol
(2R,3S,4R,5S)-1-(But-1-yl)-2-(hydroxymethyl)piperidine-3,4,5-triol

An extremely potent & selective a-D-galactosidase inhibitor. Also a selective inhibitor of GlcCer synthesis.

Vzorec: C₁₀H₂₁NO₄
Bod tání: 123…124 °C
Storage Temperature: Freezer
MDL Number: MFCD00269939
CAS číslo: 141206-42-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR