2-Butylpyrrolidine 95%
Dodavatel: Apollo Scientific
OR904755-1G
OR904755-250MG
APOSOR904755-1GEA
12400
CZK
APOSOR904755-1G
APOSOR904755-250MG
2-Butylpyrrolidine 95%
2-Butylpyrrolidine
Vzorec:
C₈H₁₇N MW: 127,227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02663449 CAS číslo: 3446-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...