Tisk…

Iron(III) p-toluenesulphonate 96%

Dodavatel: Apollo Scientific
Nebezpečí

OR904634-25G OR904634-100G OR904634-500G OR904634-5G
APOSOR904634-25GEA 827 CZK
APOSOR904634-25G APOSOR904634-100G APOSOR904634-500G APOSOR904634-5G
Iron(III) p-toluenesulphonate 96%
Iron(III) p-toluenesulphonate
Vzorec: (C₇H₇O₃S)₃·Fe
Storage Temperature: Ambient
MDL Number: MFCD01766290
CAS číslo: 77214-82-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR