5-Nitro-2-(1H-pyrrol-1-yl)pyridine 95%
Dodavatel: Apollo Scientific
Synonyms:
WAY-634964
OR904544-1G
OR904544-250MG
APOSOR904544-1GEA
28730
CZK
APOSOR904544-1G
APOSOR904544-250MG
5-Nitro-2-(1H-pyrrol-1-yl)pyridine 95%
5-Nitro-2-(1H-pyrrol-1-yl)pyridine
Vzorec:
C₉H₇N₃O₂ MW: 189,171 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05269153 CAS číslo: 157842-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...