Tisk…

4,7-Dimethyl-1H-indole-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR904418-1G OR904418-250MG
APOSOR904418-1GEA 19440 CZK
APOSOR904418-1G APOSOR904418-250MG
4,7-Dimethyl-1H-indole-2-carboxylic acid 95%
4,7-Dimethyl-1H-indole-2-carboxylic acid
Vzorec: C₁₁H₁₁NO₂
MW: 189,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664423
CAS číslo: 103988-96-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR