Tisk…

(2S)-2-(1H-Pyrrol-1-yl)propanoic acid 95%

Dodavatel: Apollo Scientific

OR904374-1G OR904374-250MG
APOSOR904374-1GEA 24600 CZK
APOSOR904374-1G APOSOR904374-250MG
(2S)-2-(1H-Pyrrol-1-yl)propanoic acid 95%
(2S)-2-(1H-Pyrrol-1-yl)propanoic acid
Vzorec: C₇H₉NO₂
MW: 139,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318697
CAS číslo: 116838-52-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR