4-Cyano-3-ethoxyphenylboronic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
(4-Cyano-3-ethoxyphenyl)boronic acid
OR904323-1G
OR904323-250MG
APOSOR904323-1GEA
6390
CZK
APOSOR904323-1G
APOSOR904323-250MG
4-Cyano-3-ethoxyphenylboronic acid 98%
4-Cyano-3-ethoxyphenylboronic acid
Vzorec:
C₉H₁₀BNO₃ MW: 190,992 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19684099 CAS číslo: 2096338-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...