4-Methyl-2-(1H-pyrrol-1-yl)-1,3-thiazole 95%
Dodavatel: Apollo Scientific
OR904301-100MG
OR904301-1G
OR904301-250MG
APOSOR904301-100MGEA
4770
CZK
APOSOR904301-100MG
APOSOR904301-1G
APOSOR904301-250MG
4-Methyl-2-(1H-pyrrol-1-yl)-1,3-thiazole 95%
4-Methyl-2-(1H-pyrrol-1-yl)-1,3-thiazole
Vzorec:
C₈H₈N₂S Storage Temperature: Ambient |
MDL Number:
MFCD02665526 CAS číslo: 383142-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...