4-Chloro-1H-pyrazole-5-carboxylic acid 96%
Dodavatel: Apollo Scientific
OR904216-1G
OR904216-5G
APOSOR904216-1GEA
1950
CZK
APOSOR904216-1G
APOSOR904216-5G
4-Chloro-1H-pyrazole-5-carboxylic acid 96%
4-Chloro-1H-pyrazole-5-carboxylic acid
Vzorec:
C₄H₃ClN₂O₂ MW: 146,532 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00466338 CAS číslo: 84547-87-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...