Tisk…

1-Pyridin-3-yl-1H-pyrrole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR904181-1G OR904181-250MG
APOSOR904181-1GEA 19440 CZK
APOSOR904181-1G APOSOR904181-250MG
1-Pyridin-3-yl-1H-pyrrole-2-carbaldehyde 95%
1-Pyridin-3-yl-1H-pyrrole-2-carbaldehyde
Vzorec: C₁₀H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD02664910
CAS číslo: 383136-42-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR