5-Cyanoquinoline 95%
Dodavatel: Apollo Scientific
OR904029-100MG
OR904029-1G
OR904029-250MG
APOSOR904029-100MGEA
4770
CZK
APOSOR904029-100MG
APOSOR904029-1G
APOSOR904029-250MG
5-Cyanoquinoline 95%
5-Cyanoquinoline
Vzorec:
C₁₀H₆N₂ MW: 154.168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02752660 CAS číslo: 59551-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...