Tisk…

(2-Formyl-1H-pyrrol-1-yl)acetic acid 95%

Dodavatel: Apollo Scientific

OR903938-1G OR903938-250MG
APOSOR903938-1GEA 12300 CZK
APOSOR903938-1G APOSOR903938-250MG
(2-Formyl-1H-pyrrol-1-yl)acetic acid 95%
(2-Formyl-1H-pyrrol-1-yl)acetic acid
Vzorec: C₇H₇NO₃
MW: 153,135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752783
CAS číslo: 67451-42-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR