N-Acetyl-2-piperidone 95%
Dodavatel: Apollo Scientific
OR903928-1G
OR903928-250MG
APOSOR903928-1GEA
9330
CZK
APOSOR903928-1G
APOSOR903928-250MG
N-Acetyl-2-piperidone 95%
N-Acetyl-2-piperidone
Vzorec:
C₇H₁₁NO₂ MW: 141,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09800363 CAS číslo: 3326-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...