Tisk…

2-Amino-4-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 95%

Dodavatel: Apollo Scientific

OR903536-100MG OR903536-1G OR903536-250MG
APOSOR903536-100MGEA 4770 CZK
APOSOR903536-100MG APOSOR903536-1G APOSOR903536-250MG
2-Amino-4-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile 95%
2-Amino-4-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Vzorec: C₁₀H₁₂N₂S
MW: 192,281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752918
CAS číslo: 60598-69-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR