Tisk…

3-Methoxy-2,2-dimethyl-3-oxopropanoic acid 98%

Dodavatel: Apollo Scientific

OR903534-5G
APOSOR903534-5GEA 4800 CZK
APOSOR903534-5G
3-Methoxy-2,2-dimethyl-3-oxopropanoic acid 98%
3-Methoxy-2,2-dimethyl-3-oxopropanoic acid
Vzorec: C₆H₁₀O₄
MW: 146,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09833517
CAS číslo: 13051-21-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR