N-(2-Cyanophenyl)acetamide 97%
Dodavatel: Apollo Scientific
OR903353-1G
OR903353-250MG
APOSOR903353-1GEA
16960
CZK
APOSOR903353-1G
APOSOR903353-250MG
N-(2-Cyanophenyl)acetamide 97%
N-(2-Cyanophenyl)acetamide
Vzorec:
C₉H₈N₂O MW: 160.173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00222087 CAS číslo: 25116-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...