Ethyl 2-(4-bromoanilino)acetate 97%
Dodavatel: Apollo Scientific
OR903261-1G
OR903261-5G
APOSOR903261-1GEA
4070
CZK
APOSOR903261-1G
APOSOR903261-5G
Ethyl 2-(4-bromoanilino)acetate 97%
Ethyl 2-(4-bromoanilino)acetate
Vzorec:
C₁₀H₁₂BrNO₂ MW: 258.112 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00461415 CAS číslo: 2521-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...